Gene Transcripts
- Gene Symbol
- Tram1
- Entrez Gene ID
- 72265
- Gene Name
- translocating chain-associating membrane protein 1
- Gene Aliases
- 1810049E02Rik, TRAMP
- Chromosome Location
- Chr. 1: 13564693 - 13589910 on Build GRCm38
- UniGene ID
- Mm.28765
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK028304.1 |
- |
4, 5 |
570 |
|
AK088814.1 |
- |
4, 5 |
561 |
|
AK142658.1 |
- |
4, 5 |
561 |
|
AK145291.1 |
- |
4, 5 |
556 |
|
AK146207.1 |
- |
4, 5 |
560 |
|
AK150551.1 |
- |
4, 5 |
567 |
|
AK151240.1 |
- |
4, 5 |
524 |
|
AK151409.1 |
- |
4, 5 |
567 |
|
AK151664.1 |
- |
4, 5 |
567 |
|
AK152338.1 |
- |
4, 5 |
567 |
|
AK152343.1 |
- |
4, 5 |
526 |
|
AK153379.1 |
- |
4, 5 |
567 |
|
AK155905.1 |
- |
4, 5 |
561 |
|
AK159203.1 |
- |
4, 5 |
561 |
|
AK159350.1 |
- |
4, 5 |
561 |
|
AK159878.1 |
- |
4, 5 |
570 |
|
AK164347.1 |
- |
4, 5 |
473 |
|
AK168708.1 |
- |
4, 5 |
503 |
|
AY029764.1 |
- |
4, 5 |
453 |
|
BC012401.1 |
- |
4, 5 |
550 |
3967323 |
CT010238.1 |
- |
4, 5 |
418 |
|
View Full Product Details