Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114328.2 | NP_001107800.1 | 8 | 1041 | |
NM_001286544.1 | NP_001273473.1 | 8 | 1041 | ||
NM_001286545.1 | NP_001273474.1 | 8 | 1041 | ||
NM_001286546.1 | NP_001273475.1 | 8 | 1027 | ||
NM_028181.5 | NP_082457.3 | 8 | 1041 | ||
GenBank mRNA | AK032645.1 | - | 8 | 913 | |
AK076325.2 | - | 8 | 968 | ||
AK128894.1 | - | 8 | 967 | ||
AK134675.1 | - | 8 | 938 | ||
BC043049.1 | - | 8 | 940 | 5704185 |