Gene Transcripts

Gene Symbol
Wdr44
Entrez Gene ID
72404
Gene Name
WD repeat domain 44
Gene Aliases
2610034K17Rik, RAB11BP, RPH11
Chromosome Location
Chr. X: 23692926 - 23806026 on Build GRCm38
UniGene ID
Mm.423
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK033628.1 - 4 432
AK139877.1 - 9 1082
BC049191.1 - 18 2501 6476607
BC068151.1 - 17 2697 30534873

View Full Product Details