Gene Transcripts

Gene Symbol
Thap1
Entrez Gene ID
73754
Gene Name
THAP domain containing, apoptosis associated protein 1
Gene Aliases
4833431A01Rik, AW490810
Chromosome Location
Chr. 8: 26158169 - 26164151 on Build GRCm38
UniGene ID
Mm.383241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199042.2 NP_950243.1 3 871
GenBank mRNA BC038639.1 - 3 827

View Full Product Details