Gene Transcripts

Gene Symbol
NAB2
Entrez Gene ID
4665
Gene Name
NGFI-A binding protein 2
Gene Aliases
MADER
Chromosome Location
Chr. 12: 57088894 - 57095476 on Build GRCh38
UniGene ID
Hs.159223
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005967.3 NP_005958.1 2 763
XM_005268894.2 XP_005268951.1 2 763
GenBank mRNA AK314229.1 - 2 543
BC007756.2 - 2 497
BC065931.1 - 2 550 6157017
U48361.1 - 2 542
X70991.1 - 1 238

View Full Product Details