Gene Transcripts
- Gene Symbol
- Gsdmcl1
- Entrez Gene ID
- 74236
- Gene Name
- gasdermin C-like 1
- Gene Aliases
- 1700022L20Rik, Gsdmdc2
- Chromosome Location
- Chr. 15: 63847309 - 63850942 on Build GRCm38
- UniGene ID
- Mm.443274
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006248.1 |
- |
5 |
891 |
|
View Full Product Details