Gene Transcripts

Gene Symbol
Spem1
Entrez Gene ID
74288
Gene Name
sperm maturation 1
Gene Aliases
1700095G12Rik
Chromosome Location
Chr. 11: 69820871 - 69822165 on Build GRCm38
UniGene ID
Mm.159159
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028855.1 NP_083131.1 2 226
GenBank mRNA BC048760.1 - 2 226
BC100505.1 - 2 218 30917608
BC144838.1 - 1 419
EF120626.1 - 2 226

View Full Product Details