Gene Transcripts

Gene Symbol
Gcc1
Entrez Gene ID
74375
Gene Name
golgi coiled coil 1
Gene Aliases
4932417P04Rik
Chromosome Location
Chr. 6: 28416603 - 28421724 on Build GRCm38
UniGene ID
Mm.332950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028900.4 NP_083176.3 1 1276
GenBank mRNA AK016531.1 - 1225
AK032253.1 - 1 1276
BC066807.1 - 1 1245 30101518

View Full Product Details