Gene Transcripts

Gene Symbol
Spata9
Entrez Gene ID
75571
Gene Name
spermatogenesis associated 9
Gene Aliases
1700030K01Rik, 4930599C08Rik, A930023H06Rik
Chromosome Location
Chr. 13: 75960003 - 75998987 on Build GRCm38
UniGene ID
Mm.443291
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029343.3 NP_083619.1 4 547
GenBank mRNA AK006554.1 - 4 548
AK020885.1 - 2 319
BC048558.1 - 4 527 6703288

View Full Product Details