Gene Transcripts

Gene Symbol
Wdr54
Entrez Gene ID
75659
Gene Name
WD repeat domain 54
Gene Aliases
1700030E05Rik
Chromosome Location
Chr. 6: 83152710 - 83156406 on Build GRCm38
UniGene ID
Mm.130665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023790.2 NP_076279.1 9, 10 1077
GenBank mRNA AF053628.1 - 9, 10 1065
AK006536.1 - 9, 10 977
AK154345.1 - 9, 10 1068
BC083332.1 - 10, 11 982 6819838
BY540708.1 - 2, 3 180

View Full Product Details