Gene Transcripts

Gene Symbol
Rab11fip1
Entrez Gene ID
75767
Gene Name
RAB11 family interacting protein 1 (class I)
Gene Aliases
2010200K21Rik, 4833414G05Rik, Rcp
Chromosome Location
Chr. 8: 27138773 - 27174646 on Build GRCm38
UniGene ID
Mm.205335
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080813.2 NP_001074282.1 1 361
NM_029423.2 NP_083699.2 1 361
GenBank mRNA AK014696.1 - 1 343
AK030769.1 - 1 364
AK086797.1 - 1 359
AK089792.1 - 1 343
AK151024.1 - 1 362
AK166735.1 - 1 356
BB619642.1 - 1 362
BC125400.1 - 1 346 40129915
BC132094.1 - 1 346 40130371

View Full Product Details