Gene Transcripts

Gene Symbol
Cyp2d26
Entrez Gene ID
76279
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 26
Gene Aliases
1300006E06Rik
Chromosome Location
Chr. 15: 82790107 - 82794452 on Build GRCm38
UniGene ID
Mm.439931
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029562.2 NP_083838.1 7 1156
GenBank mRNA AK004915.1 - 7 1157
BC023241.1 - 7 1145 4222538

View Full Product Details