Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135100.2 | NP_001128572.1 | 4 | 779 | |
NM_029646.3 | NP_083922.1 | 4 | 779 | ||
GenBank mRNA | AK008082.1 | - | 4 | 727 | |
BC016254.1 | - | 4 | 758 | 4912097 | |
EU636995.1 | - | 3 | 196 | ||
EU636996.1 | - | 3 | 196 |