Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030083.2 | NP_084359.1 | 2 | 261 | |
GenBank mRNA | AK018782.1 | - | 2 | 262 | |
BC038659.1 | - | 2 | 215 | ||
BC048499.1 | - | 2 | 267 | 6583055 | |
BC062267.1 | - | 2 | 231 | 1532489 |