Gene Transcripts
- Gene Symbol
- Krit1
- Entrez Gene ID
- 79264
- Gene Name
- KRIT1, ankyrin repeat containing
- Gene Aliases
- 2010007K12Rik, A630036P20Rik, AA432855, AI450393, AI643869, BB155247, BB235701, Ccm1
- Chromosome Location
- Chr. 5: 3803165 - 3844515 on Build GRCm38
- UniGene ID
- Mm.32368
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170552.1 |
NP_001164023.1
|
5, 6 |
855 |
|
NM_030675.3 |
NP_109600.2
|
5, 6 |
809 |
|
GenBank mRNA |
AF306509.1 |
- |
5, 6 |
522 |
|
AF310134.1 |
- |
5, 6 |
785 |
|
AK039264.1 |
- |
|
830 |
|
AK085107.1 |
- |
5, 6 |
837 |
|
AK144667.1 |
- |
5, 6 |
791 |
|
AY328895.1 |
- |
4, 5 |
491 |
|
AY464945.1 |
- |
4, 5 |
491 |
|
BC016087.1 |
- |
5, 6 |
759 |
|
BC044820.1 |
- |
4, 5 |
550 |
|
BC054819.1 |
- |
5, 6 |
900 |
|
BC085252.1 |
- |
5, 6 |
615 |
|
BC141042.1 |
- |
5, 6 |
653 |
9055654 |
View Full Product Details