Gene Transcripts

Gene Symbol
Rilpl2
Entrez Gene ID
80291
Gene Name
Rab interacting lysosomal protein-like 2
Gene Aliases
Chromosome Location
Chr. 5: 124463265 - 124478235 on Build GRCm38
UniGene ID
Mm.425280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030259.1 NP_084535.1 3 700
GenBank mRNA AK133760.1 - 3 832
AK150123.1 - 3 804
AK150252.1 - 3 803
BC003324.1 - 3 700 3156139

View Full Product Details