Gene Transcripts

Gene Symbol
NNMT
Entrez Gene ID
4837
Gene Name
nicotinamide N-methyltransferase
Gene Aliases
Chromosome Location
Chr. 11: 114295813 - 114312516 on Build GRCh38
UniGene ID
Hs.503911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006169.2 NP_006160.1 1 805
GenBank mRNA AA402535.1 - 1 551
AK026845.1 - 1 181
BC000234.1 - 3 325 3350358
CR541907.1 - 1 61
U08021.1 - 1 178

View Full Product Details