Gene Transcripts

Gene Symbol
Lactb
Entrez Gene ID
80907
Gene Name
lactamase, beta
Gene Aliases
LACT-1, Lact1, Mrpl56
Chromosome Location
Chr. 9: 66955393 - 66975484 on Build GRCm38
UniGene ID
Mm.157882
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030717.1 NP_109642.1 5 1145
GenBank mRNA AF317900.1 - 5 1145
BC046293.1 - 5 1072 6484767
BC147809.1 - 5 1090 9087526

View Full Product Details