Gene Transcripts

Gene Symbol
Sil1
Entrez Gene ID
81500
Gene Name
endoplasmic reticulum chaperone SIL1 homolog (S. cerevisiae)
Gene Aliases
1810057E01Rik, AI042831, wz
Chromosome Location
Chr. 18: 35266396 - 35499052 on Build GRCm38
UniGene ID
Mm.291482
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030749.2 NP_109674.2 7 844
GenBank mRNA AJ297884.1 - 7 785
AK170415.1 - 7 797
AK192274.1 - 2 164
AK219586.1 - 2 164
BC016119.1 - 7 821 4489217
BC016466.1 - 7 807 3582798
CO428760.1 - 4 387

View Full Product Details