Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031186.3 | NP_112463.2 | 1 | 361 | |
GenBank mRNA | AF221095.1 | - | 1 | 360 | |
AK141945.1 | - | 2 | 510 | ||
BC079622.1 | - | 1 | 276 | 30535990 | |
BC112404.1 | - | 1 | 273 | 40060424 |