Gene Transcripts

Gene Symbol
Smarcd1
Entrez Gene ID
83797
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1
Gene Aliases
AA407987, Baf60a, D15Kz1
Chromosome Location
Chr. 15: 99702287 - 99713995 on Build GRCm38
UniGene ID
Mm.273756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031842.2 NP_114030.2 4, 5 567
GenBank mRNA BC026783.1 - 4, 5 479
BC059921.1 - 4, 5 537
BC079839.1 - 4, 5 513 6810462
CV562378.1 - 4, 5 567
M25773.1 - 4, 5 523
U66620.1 - 4, 5 523

View Full Product Details