Gene Transcripts
- Gene Symbol
- Dnaja3
- Entrez Gene ID
- 83945
- Gene Name
- DnaJ (Hsp40) homolog, subfamily A, member 3
- Gene Aliases
- 1200003J13Rik, 1810053A11Rik, C81173, Tid-1, Tid1l
- Chromosome Location
- Chr. 16: 4684070 - 4707695 on Build GRCm38
- UniGene ID
- Mm.248337
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135112.1 |
NP_001128584.1
|
2, 3 |
402 |
|
NM_023646.4 |
NP_076135.3
|
2, 3 |
402 |
|
GenBank mRNA |
AF325535.1 |
- |
2, 3 |
443 |
|
AF326358.1 |
- |
2, 3 |
443 |
|
AK004575.1 |
- |
2, 3 |
352 |
|
AK004910.1 |
- |
2, 3 |
354 |
|
AK007852.1 |
- |
2, 3 |
343 |
|
AK031250.1 |
- |
2, 3 |
359 |
|
AK167511.1 |
- |
2, 3 |
364 |
|
AK214348.1 |
- |
1, 2 |
135 |
|
AY009320.1 |
- |
2, 3 |
443 |
|
BC003920.1 |
- |
2, 3 |
335 |
|
BC027240.1 |
- |
2, 3 |
364 |
2654866 |
CJ148764.1 |
- |
2 |
403 |
|
View Full Product Details