Gene Transcripts

Gene Symbol
Mmp1b
Entrez Gene ID
83996
Gene Name
matrix metallopeptidase 1b (interstitial collagenase)
Gene Aliases
Mcol-B, Mcolb
Chromosome Location
Chr. 9: 7367640 - 7388026 on Build GRCm38
UniGene ID
Mm.379271
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032007.3 NP_114396.3 4 636
GenBank mRNA AJ278461.1 - 4 605

View Full Product Details