Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289706.1 | NP_001276635.1 | 7 | 721 | |
NM_001289707.1 | NP_001276636.1 | 6 | 646 | ||
NM_001289708.1 | NP_001276637.1 | 5 | 563 | ||
NM_020257.2 | NP_064653.1 | 6 | 634 | ||
GenBank mRNA | AF121352.1 | - | 6 | 607 | |
AF350411.1 | - | 6 | 611 | ||
AK041724.1 | - | 5 | 1349 | ||
AY137344.1 | - | 6 | 645 | ||
AY137345.1 | - | 6 | 633 | ||
AY256574.1 | - | 7 | 712 | ||
AY256575.1 | - | 6 | 635 | ||
AY256576.1 | - | 5 | 552 | ||
BC139192.1 | - | 6 | 607 | 8862214 | |
BC139193.1 | - | 6 | 607 | 8862215 | |
BC145574.1 | - | 6 | 619 | 9054140 | |
HQ713440.1 | - | 5 | 541 |