Gene Transcripts

Gene Symbol
Glce
Entrez Gene ID
93683
Gene Name
glucuronyl C5-epimerase
Gene Aliases
1110017N23Rik, AI747411, C130034A12Rik, Hsepi
Chromosome Location
Chr. 9: 62057248 - 62122720 on Build GRCm38
UniGene ID
Mm.24411
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033320.4 NP_201577.3 1 476
GenBank mRNA AF325532.1 - 3 701
AF330049.1 - 1 710
BC094587.1 - 1 476 30618522

View Full Product Details