Gene Transcripts

Gene Symbol
Pcdhb3
Entrez Gene ID
93874
Gene Name
protocadherin beta 3
Gene Aliases
PcdhbC
Chromosome Location
Chr. 18: 37300799 - 37304585 on Build GRCm38
UniGene ID
Mm.82308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053128.2 NP_444358.2 1 1200
GenBank mRNA AF326296.1 - 1 1016
AK030570.1 - 1 1200
AY013785.1 - 1 1016
BC079557.1 - 1 1114 30360109

View Full Product Details