Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052976.3 | NP_443208.1 | 3 | 860 | |
GenBank mRNA | AK028822.1 | - | 3 | 344 | |
AK031419.1 | - | 3 | 853 | ||
AK036038.1 | - | 3 | 861 | ||
AK039304.1 | - | 2 | 1028 | ||
AK043239.1 | - | 3 | 861 | ||
AK046214.1 | - | 3 | 316 | ||
AK048153.1 | - | 3 | 861 | ||
AK077982.1 | - | 3 | 485 | ||
AK082551.1 | - | 3 | 500 | ||
AK087469.1 | - | 3 | 324 | ||
BC004845.1 | - | 4 | 458 | 2650009 |