Gene Transcripts

Gene Symbol
Myl9
Entrez Gene ID
98932
Gene Name
myosin, light polypeptide 9, regulatory
Gene Aliases
AI327049, MLC20, Mylc2c, RLC-C
Chromosome Location
Chr. 2: 156775435 - 156781658 on Build GRCm38
UniGene ID
Mm.271770
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172118.1 NP_742116.1 2 194
GenBank mRNA AK007972.1 - 2 204
AK020258.1 - 2 195
AK159880.1 - 2 239
BC049974.1 - 2 229 4912899

View Full Product Details