Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006179.4 | NP_006170.1 | 2 | 919 | |
XM_005258962.3 | XP_005259019.1 | 3 | 1040 | ||
XM_006723232.3 | XP_006723295.1 | 3 | 1082 | ||
XM_011527008.2 | XP_011525310.1 | 2 | 823 | ||
XM_011527009.2 | XP_011525311.1 | 2 | 1912 | ||
XM_011527010.2 | XP_011525312.1 | 2 | 2030 | ||
GenBank mRNA | BC012421.2 | - | 2 | 919 |