Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133952.2 | NP_598713.2 | 2, 3 | 321 | |
GenBank mRNA | AK028912.1 | - | 2, 3 | 307 | |
AK047292.1 | - | 2, 3 | 268 | ||
AK047476.1 | - | 2, 3 | 268 | ||
AK088034.1 | - | 2, 3 | 273 | ||
AK166809.1 | - | 2, 3 | 304 | ||
AK202434.1 | - | 1, 2 | 96 | ||
BB842393.1 | - | 2, 3 | 316 | ||
BC004717.1 | - | 2, 3 | 310 | 3582116 |