Gene Transcripts

Gene Symbol
Acad11
Entrez Gene ID
102632
Gene Name
acyl-Coenzyme A dehydrogenase family, member 11
Gene Aliases
5730439E10Rik, AI987948
Chromosome Location
Chr. 9: 104063703 - 104127646 on Build GRCm38
UniGene ID
Mm.269254
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175324.3 NP_780533.2 3 392
GenBank mRNA AK077531.1 - 3 393
AK128998.1 - 3 371
AK139918.1 - 3 335
AK145561.1 - 3 381
AK145834.1 - 3 372
AK216697.1 - 2 119
BC045199.1 - 3 356 5133197

View Full Product Details