Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139142.2 | NP_631881.1 | 11 | 1698 | |
GenBank mRNA | AJ428067.1 | - | 1711 | ||
AJ964961.1 | - | 11 | 1672 | ||
AK043297.1 | - | 11 | 1709 | ||
AK080979.1 | - | 1711 | |||
AK140445.1 | - | 1709 | |||
AK160468.1 | - | 1709 | |||
BC118933.2 | - | 11 | 1698 | 40090800 |