Gene Transcripts

Gene Symbol
Tmed4
Entrez Gene ID
103694
Gene Name
transmembrane emp24 protein transport domain containing 4
Gene Aliases
1110014L17Rik, AI326346
Chromosome Location
Chr. 11: 6270714 - 6274837 on Build GRCm38
UniGene ID
Mm.254495
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134020.1 NP_598781.1 1 94
GenBank mRNA AF368921.1 - 1 125
AK078425.1 - 1 105
AK158876.1 - 1 127
BC023041.1 - 1 94 5360236

View Full Product Details