Gene Transcripts

Gene Symbol
Rhox9
Entrez Gene ID
104384
Gene Name
reproductive homeobox 9
Gene Aliases
1600026O01Rik, Gpbox, Psx2
Chromosome Location
Chr. X: 37899097 - 37901770 on Build GRCm38
UniGene ID
Mm.156958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023894.1 NP_076383.1 4 841
GenBank mRNA AF201698.1 - 4 841
AK018818.1 - 3 282
BC099451.1 - 4 780 30716051
BC099963.1 - 2 2381 30083999
DQ058646.1 - 4 841

View Full Product Details