Gene Transcripts

Gene Symbol
ACVR2B
Entrez Gene ID
93
Gene Name
activin A receptor type 2B
Gene Aliases
ACTRIIB, ActR-IIB, HTX4
Chromosome Location
Chr. 3: 38454299 - 38493142 on Build GRCh38
UniGene ID
Hs.174273
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106.3 NP_001097.2 6 793
XM_005265583.3 XP_005265640.1 6 3255
XM_017007514.1 XP_016863003.1 6 3234
XM_017007515.1 XP_016863004.1 6 879
XM_017007516.1 XP_016863005.1 6 4203
GenBank mRNA AB209449.1 - 5 4558
BC096243.1 - 6 773 40005760
BC096244.1 - 6 773 40005761
BC096245.1 - 5 855 40005763
BC099642.4 - 6 773 40005762
X77533.1 - 6 773

View Full Product Details