Gene Transcripts

Gene Symbol
OR3A1
Entrez Gene ID
4994
Gene Name
olfactory receptor family 3 subfamily A member 1
Gene Aliases
OLFRA03, OR17-40, OR17-82, OR40
Chromosome Location
Chr. 17: 3288318 - 3336087 on Build GRCh38
UniGene ID
Hs.647410
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002550.2 NP_002541.2 1 770
XM_017024695.1 XP_016880184.1 2 861
XM_017024696.1 XP_016880185.1 2 835
GenBank mRNA BC069414.1 - 1 770
BC096187.3 - 1 770 40004009
BC096188.3 - 1 770 40004010
BC096189.3 - 1 770 40004011
BC096190.1 - 1 770 40004012

View Full Product Details