Gene Transcripts

Gene Symbol
Cops8
Entrez Gene ID
108679
Gene Name
COP9 (constitutive photomorphogenic) homolog, subunit 8 (Arabidopsis thaliana)
Gene Aliases
9430009J09Rik, AA408242, Csn8, Sgn8
Chromosome Location
Chr. 1: 90603425 - 90613341 on Build GRCm38
UniGene ID
Mm.119177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133805.3 NP_598566.3 4 396
GenBank mRNA AF482000.1 - 4 857
AK079111.1 - 4 330
AK079522.1 - 4 344
AK088323.1 - 4 316
AK090013.1 - 4 325
AK142989.1 - 3 1365
AK145899.1 - 4 323
AK162502.1 - 4 331

View Full Product Details