Gene Transcripts
- Gene Symbol
- Pfdn4
- Entrez Gene ID
- 109054
- Gene Name
- prefoldin 4
- Gene Aliases
- 0610025M10Rik, 1810063B06Rik, C-1, C1, Pfd4
- Chromosome Location
- Chr. 2: 170496428 - 170519070 on Build GRCm38
- UniGene ID
- Mm.28808
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001013369.2 |
NP_001013387.1
|
3, 4 |
413 |
|
NM_001110152.2 |
NP_001103622.1
|
3, 4 |
481 |
|
NM_001199902.1 |
NP_001186831.1
|
3, 4 |
469 |
|
GenBank mRNA |
AK007940.1 |
- |
3, 4 |
274 |
|
AK136249.1 |
- |
3, 4 |
482 |
|
BC048510.1 |
- |
3, 4 |
253 |
|
BC065407.1 |
- |
3, 4 |
413 |
|
BC147673.1 |
- |
3, 4 |
314 |
9056646 |
BC147674.1 |
- |
3, 4 |
314 |
9056647 |
BU503885.1 |
- |
2, 3 |
252 |
|
CB523981.1 |
- |
1, 2 |
89 |
|
View Full Product Details