Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_176843.3 | NP_789813.1 | 2 | 1306 | |
GenBank mRNA | AK143868.1 | - | 2 | 1269 | |
AK144770.1 | - | 2 | 1281 | ||
AK155499.1 | - | 2 | 1268 | ||
AK166149.1 | - | 2 | 1279 | ||
AK168795.1 | - | 2 | 1279 | ||
AK170584.1 | - | 2 | 1268 | ||
BC039214.1 | - | 2 | 1288 | 4191047 |