Gene Transcripts

Gene Symbol
Sri
Entrez Gene ID
109552
Gene Name
sorcin
Gene Aliases
2210417O06Rik, 2900070H08Rik, Sor
Chromosome Location
Chr. 5: 8046078 - 8069314 on Build GRCm38
UniGene ID
Mm.96211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080974.2 NP_001074443.1 4 282
NM_025618.3 NP_079894.2 4 322
GenBank mRNA AK008404.1 - 4 274
AK008783.1 - 4 323
AK008970.1 - 4 271
AK013762.1 - 1 1148
AK134747.1 - 4 284
AK146071.1 - 4 279
AK146911.1 - 4 263
AK168572.1 - 4 263
AK168792.1 - 4 263
AK168900.1 - 4 254
BC065790.1 - 4 249 948616
BY708689.1 - 4 320

View Full Product Details