Gene Transcripts

Gene Symbol
Lmo1
Entrez Gene ID
109594
Gene Name
LIM domain only 1
Gene Aliases
Rbtn-1, Rbtn1, Ttg1
Chromosome Location
Chr. 7: 109138565 - 109175343 on Build GRCm38
UniGene ID
Mm.360145
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AA050915.1 - 2 256
AK133586.1 - 2 145
AW494084.1 - 2 339
BC053074.1 - 2 395 5693266
DV662082.1 - 2 606

View Full Product Details