Gene Transcripts

Gene Symbol
P2RY6
Entrez Gene ID
5031
Gene Name
pyrimidinergic receptor P2Y6
Gene Aliases
P2Y6
Chromosome Location
Chr. 11: 73264503 - 73298625 on Build GRCh38
UniGene ID
Hs.16362
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277204.1 NP_001264133.1 3 1388
NM_001277205.1 NP_001264134.1 4 1335
NM_001277206.1 NP_001264135.1 3 1293
NM_001277207.1 NP_001264136.1 3 1235
NM_001277208.1 NP_001264137.1 2 1302
NM_176796.2 NP_789766.1 4 1368
NM_176797.2 NP_789767.1 3 1208
NM_176798.2 NP_789768.1 4 1485
XM_005274022.3 XP_005274079.1 2 3476
XM_006718571.3 XP_006718634.1 3 3561
XM_011545076.2 XP_011543378.1 3 2691
XM_011545077.2 XP_011543379.1 4 2810
XM_011545079.2 XP_011543381.1 2 1124
GenBank mRNA AF007891.1 - 3 1174
AF007892.1 - 4 1485
AF007893.1 - 3 2009
AF218005.1 - 3 1245
AF498920.1 - 1 943
AX775903.1 - 4 1485
BC000571.2 - 2 1183 3162734
BC009391.2 - 2 1242 4128941
BT006771.1 - 1 943
U52464.1 - 3 1146
X97058.1 - 3 1219

View Full Product Details