Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000917.3 | NP_000908.2 | 7 | 1102 | |
NM_001017962.2 | NP_001017962.1 | 7 | 1102 | ||
NM_001142595.1 | NP_001136067.1 | 8 | 1195 | ||
NM_001142596.1 | NP_001136068.1 | 7 | 1102 | ||
GenBank mRNA | AK222960.1 | - | 7 | 990 | |
AK312593.1 | - | 7 | 988 | ||
BC034998.1 | - | 7 | 995 | 4797051 | |
CD013929.1 | - | 7 | 981 | ||
CD013930.1 | - | 8 | 1074 | ||
M24486.1 | - | 7 | 979 | ||
M24487.1 | - | 7 | 979 |