Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321781.1 | NP_001308710.1 | 2 | 167 | |
NM_001321782.1 | NP_001308711.1 | 2 | 160 | ||
NM_152516.3 | NP_689729.1 | 2 | 264 | ||
XM_011532558.2 | XP_011530860.1 | 2 | 262 | ||
XM_011532559.2 | XP_011530861.1 | 2 | 215 | ||
XM_017003412.1 | XP_016858901.1 | 2 | 72 | ||
GenBank mRNA | AB178811.1 | - | 2 | 248 | |
AK223607.1 | - | 2 | 253 | ||
AK294203.1 | - | 2 | 232 | ||
BC009266.2 | - | 2 | 217 | ||
BC022046.1 | - | 2 | 236 | 4046433 | |
D85433.1 | - | 2 | 164 | ||
DT217730.1 | - | 2 | 166 | ||
HY001060.1 | - | 2 | 264 | ||
HY039942.1 | - | 2 | 167 |