Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_002228.3 | NP_002219.1 |
GenBank mRNA | BC002646.2 | - |
BC006175.1 | - | |
BC009874.2 | - | |
BC068522.1 | - | |
BG831943.1 | - | |
BI793062.1 | - | |
BT019759.1 | - | |
BU782554.1 | - | |
CD370800.1 | - | |
CR541724.1 | - | |
CR541752.1 | - | |
ENST00000371222.0 | - |