Gene Transcripts

Gene Symbol
Tram2
Entrez Gene ID
170829
Gene Name
translocating chain-associating membrane protein 2
Gene Aliases
C330003D03Rik
Chromosome Location
Chr. 1: 21001397 - 21079225 on Build GRCm38
UniGene ID
Mm.185680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177409.3 NP_803128.1 3, 4 399
GenBank mRNA AK082751.1 - 3, 4 400
AY029530.1 - 3, 4 336
BC018212.1 - 3, 4 384 3980004

View Full Product Details