Gene Transcripts
- Gene Symbol
- Slc24a5
- Entrez Gene ID
- 317750
- Gene Name
- solute carrier family 24, member 5
- Gene Aliases
- F630045L20Rik, JSX, NCKX5, NCX5, Oca6
- Chromosome Location
- Chr. 2: 125068127 - 125088677 on Build GRCm38
- UniGene ID
- Mm.330538
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB085629.1 |
- |
1, 2 |
186 |
|
View Full Product Details