Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr. 10: 91923769 - 92030998 on Build GRCh38
UniGene ID
Hs.500526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003972.2 NP_003963.1 5 528
XM_011540326.1 XP_011538628.1 5 738
XM_011540327.2 XP_011538629.1 2 293
XM_011540328.2 XP_011538630.1 2 121
XM_011540329.2 XP_011538631.1 5 738
XM_017016877.1 XP_016872366.1 4 590
XM_017016878.1 XP_016872367.1 4 416
XM_017016879.1 XP_016872368.1 4 416
XM_017016880.1 XP_016872369.1 4 416
XM_017016881.1 XP_016872370.1 3 269
GenBank mRNA AF038362.1 - 5 547
AJ001017.2 - 5 442
AK308399.1 - 5 738
BC112201.1 - 5 512 8327669

View Full Product Details