Gene Transcripts
- Gene Symbol
- MTF2
- Entrez Gene ID
- 22823
- Gene Name
- metal response element binding transcription factor 2
- Gene Aliases
- M96, PCL2, TDRD19A, dJ976O13.2
- Chromosome Location
- Chr. 1: 93079219 - 93139081 on Build GRCh38
- UniGene ID
- Hs.31016
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ010014.1 |
- |
15 |
2099 |
|
View Full Product Details