Gene Transcripts

Gene Symbol
TCF4
Entrez Gene ID
6925
Gene Name
transcription factor 4
Gene Aliases
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location
Chr. 18: 55222331 - 55635993 on Build GRCh38
UniGene ID
Hs.605153
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083962.1 NP_001077431.1 15 1771
NM_001243226.2 NP_001230155.2 16 1561
NM_001243227.1 NP_001230156.1 14 1605
NM_001243228.1 NP_001230157.1 15 1715
NM_001243230.1 NP_001230159.1 14 1244
NM_001243231.1 NP_001230160.1 13 1205
NM_001243232.1 NP_001230161.1 11 1227
NM_001243233.1 NP_001230162.1 11 1168
NM_001243234.1 NP_001230163.1 8 800
NM_001243235.1 NP_001230164.1 8 800
NM_001243236.1 NP_001230165.1 8 816
NM_001306207.1 NP_001293136.1 14 1396
NM_001306208.1 NP_001293137.1 11 1227
NM_003199.2 NP_003190.1 15 1771
XM_005266739.3 XP_005266796.2 14 1411
XM_005266741.3 XP_005266798.1 15 1371
XM_005266743.4 XP_005266800.1 14 1232
XM_005266744.3 XP_005266801.1 14 1352
XM_005266745.3 XP_005266802.1 14 1214
XM_005266747.3 XP_005266804.1 13 1208
XM_005266749.3 XP_005266806.1 11 999
XM_005266752.4 XP_005266809.1 11 1569
XM_005266754.3 XP_005266811.1 10 993
XM_005266755.4 XP_005266812.1 11 1028
XM_005266761.3 XP_005266818.1 8 1309
XM_006722536.2 XP_006722599.1 15 1694
XM_006722537.2 XP_006722600.1 15 1327
XM_006722538.2 XP_006722601.1 14 1349
XM_011526158.2 XP_011524460.1 14 2325
XM_011526160.2 XP_011524462.1 10 1378
XM_017025934.1 XP_016881423.1 14 1408
XM_017025935.1 XP_016881424.1 14 1408
XM_017025936.1 XP_016881425.1 14 1411
XM_017025937.1 XP_016881426.1 14 1165
XM_017025938.1 XP_016881427.1 15 1324
XM_017025939.1 XP_016881428.1 15 1371
XM_017025940.1 XP_016881429.1 15 1694
XM_017025941.1 XP_016881430.1 15 1327
XM_017025942.1 XP_016881431.1 15 1371
XM_017025943.1 XP_016881432.1 15 1371
XM_017025944.1 XP_016881433.1 14 1629
XM_017025945.1 XP_016881434.1 14 1629
XM_017025946.1 XP_016881435.1 14 1646
XM_017025947.1 XP_016881436.1 14 1232
XM_017025948.1 XP_016881437.1 14 1629
XM_017025949.1 XP_016881438.1 14 1349
XM_017025950.1 XP_016881439.1 14 1626
XM_017025951.1 XP_016881440.1 13 1205
XM_017025952.1 XP_016881441.1 11 1219
XM_017025953.1 XP_016881442.1 11 999
XM_017025954.1 XP_016881443.1 11 1580
XM_017025955.1 XP_016881444.1 10 1314
XM_017025956.1 XP_016881445.1 10 993
XM_017025957.1 XP_016881446.1 8 1309
GenBank mRNA AB209741.1 - 10 879
AK095041.1 - 11 1227
AK096862.1 - 13 1205
AK122765.1 - 15 1771
AK299169.1 - 14 1605
AK300038.1 - 8 746
AK300612.1 - 11 1168
AK300636.1 - 10 898
AK301144.1 - 14 1382
AK315074.1 - 16 1703
AK316165.1 - 10 942
AL518903.3 - 8 816
BC031056.1 - 16 1522
BC125084.1 - 14 1185 40117795
BC125085.1 - 14 1185 40117798
FR748210.1 - 14 1159
FR748211.1 - 14 1159
FR748212.1 - 12 979
FR748213.1 - 12 979
FR748214.1 - 13 1087
FR748215.1 - 11 907
FR748216.1 - 14 1150
FR748217.1 - 13 1033
FR748218.1 - 9 769
FR748219.1 - 11 946
FR748220.1 - 8 679
FR748221.1 - 8 679
FR748222.1 - 8 679
FR748223.1 - 6 511
M74718.1 - 8 631
M74719.1 - 15 1358
X52079.1 - 12 1024

View Full Product Details